Merck
  • Validated method for the determination of piroxicam by capillary zone electrophoresis and its application to tablets.
Validated method for the determination of piroxicam by capillary zone electrophoresis and its application to tablets.
Journal of analytical methods in chemistry (2014-10-09)
Arın Gül Dal, Zeynep Oktayer, Dilek Doğrukol-Ak
ABSTRACT
Simple and rapid capillary zone electrophoretic method was developed and validated in this study for the determination of piroxicam in tablets. The separation of piroxicam was conducted in a fused-silica capillary by using 10 mM borate buffer (pH 9.0) containing 10% (v/v) methanol as background electrolyte. The optimum conditions determined were 25 kV for separation voltage and 1 s for injection time. Analysis was carried out with UV detection at 204 nm. Naproxen sodium was used as an internal standard. The method was linear over the range of 0.23-28.79 µg/mL. The accuracy and precision were found to be satisfied within the acceptable limits (<2%). The LOD and LOQ were found to be 0.07 and 0.19 µg/mL, respectively. The method described here was applied to tablet dosage forms and the content of a tablet was found in the limits of USP-24 suggestions. To compare the results of capillary electrophoretic method, UV spectrophotometric method was developed and the difference between two methods was found to be insignificant. The capillary zone electrophoretic method developed in this study is rapid, simple, and suitable for routine analysis of piroxicam in pharmaceutical tablets.
Sigma-Aldrich
Naproxen sodium, 98.0-102.0%
Sigma-Aldrich
Naproxen sodium, meets USP testing specifications
USP
Naproxen sodium, United States Pharmacopeia (USP) Reference Standard
Supelco
Naproxen sodium, Pharmaceutical Secondary Standard; Certified Reference Material