Skip to Content
Merck
All Photos(4)

Documents

M1275

Sigma-Aldrich

Naproxen sodium

98.0-102.0%

Synonym(s):

(S)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NaO3
CAS Number:
Molecular Weight:
252.24
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

98.0-102.0%

form

powder

solubility

water: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

originator

Bayer

SMILES string

[Na+].COc1ccc2cc(ccc2c1)[C@H](C)C([O-])=O

InChI

1S/C14H14O3.Na/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10;/h3-9H,1-2H3,(H,15,16);/q;+1/p-1/t9-;/m0./s1

InChI key

CDBRNDSHEYLDJV-FVGYRXGTSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Naproxen belongs to phenylacetic acid class of Non-steroidal anti-inflammatory drugs (NSAIDs).

Application

Naproxen sodium has been used to evaluate the thermodynamics of biomolecular interaction with bovine and human serum albumin. It has also been used to evaluate its efficiency in reducing the response of spinal dorsal horn neurons to noxious knee joint rotation.

Biochem/physiol Actions

Naproxen binds preferably to serum albumin. The S-enantiomer of naproxen is 28-fold more potential as an anti-inflammatory drug compared to the R-isomer. The R-isomer is found to be a liver toxin and causes gastrointestinal disorders.
Cyclooxygenase (Prostaglandin H synthase 1 and 2) inhibitor.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Xia et al.
Plant communications, 4(6), 100632-100632 (2023-05-31)
The phytohormone auxin plays central roles in many growth and developmental processes in plants. Development of chemical tools targeting the auxin pathway is useful for both plant biology and agriculture. Here we reveal that naproxen, a synthetic compound with anti-inflammatory
Binding thermodynamics of Diclofenac and Naproxen with human and bovine serum albumins: A calorimetric and spectroscopic study
Bou-Abdallah F, et al.
The Journal of Chemical Thermodynamics, 103, 299-309 (2016)
Arın Gül Dal et al.
Journal of analytical methods in chemistry, 2014, 352698-352698 (2014-10-09)
Simple and rapid capillary zone electrophoretic method was developed and validated in this study for the determination of piroxicam in tablets. The separation of piroxicam was conducted in a fused-silica capillary by using 10 mM borate buffer (pH 9.0) containing 10%
Flores Martín-Reyes et al.
Antioxidants (Basel, Switzerland), 12(12) (2023-12-23)
Inflammatory Bowel Diseases (IBD) are a group of chronic, inflammatory disorders of the gut. The incidence and activity of IBD are determined by both genetic and environmental factors. Among these factors, polymorphisms in genes related to autophagy and the consumption
Margarita Valero et al.
Langmuir : the ACS journal of surfaces and colloids, 26(13), 10561-10571 (2010-05-15)
The associative structures between F127 Pluronic micelles and four drugs, namely, lidocaine (LD), pentobarbital sodium salt (PB), sodium naproxen (NP), and sodium salicylate (SAL), were studied by small-angle neutron scattering (SANS). Different outcomes for the micellar aggregates are observed, which

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service