Merck
  • Dog UDP-glucuronosyltransferase enzymes of subfamily 1A: cloning, expression, and activity.
Dog UDP-glucuronosyltransferase enzymes of subfamily 1A: cloning, expression, and activity.
Drug metabolism and disposition: the biological fate of chemicals (2014-10-11)
Johanna Troberg, Erkka Järvinen, Maria Muniz, Nina Sneitz, Johanna Mosorin, Marja Hagström, Moshe Finel
ABSTRACT
Understanding drug glucuronidation in the dog, a preclinical animal, is important but currently poorly characterized at the level of individual enzymes. We have constructed cDNAs for the 10 dog UDP-glucuronosyltransferases of subfamily 1A (dUGT1As), expressed them in insect cells, and assayed their activity as well as the activity of the nine human UGT1As, toward 14 compounds. The goal was to find out whether individual dUGT1As and individual human UGT1As have similar substrate specificities. The results revealed similarities but also many differences. For example, similarly to the human UGT1A10, dUGT1A11 exhibited high glucuronidation activity toward the 3-OH of 17-β-estradiol, 17-α-estradiol, and ethinylestradiol, and also conjugated the drug entacapone. Unlike the human UGT1A10, however, it failed to catalyze considerable rates of R-propranolol, diclofenac, and indomethacin glucuronidation. The estrogen glucuronidation assays revealed that dUGT1A8 and dUGT1A10 have a capacity to catalyze the formation of (linked) diglucuronides, an activity no human UGT1A exhibited. dUGT1A2-dUGT1A4 are homologs of the human UGT1A4, but none of them catalyzed N-glucuronidation of dexmedetomidine. Contrary to the human UGT1A4, however, dUGT1A2-dUGT1A4 catalyzed indomethacin and diclofenac glucuronidation. It may be concluded that, perhaps with the exception of UGT1A6, high similarities in substrate specificity between individual dog and human UGTs of subfamily 1A are rare or partial. Activity assays with liver and intestine microsomes of both dog and human further revealed interspecies differences, particularly in glucuronidation rates. In the dog, the microsomes assays also strongly suggested important roles for dUGTs of other subfamilies, mainly in the liver.
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Testosterone, ≥98%
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Testosterone, European Pharmacopoeia (EP) Reference Standard
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
USP
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
Ethinylestradiol, British Pharmacopoeia (BP) Assay Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Magnesium chloride hexahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC