Skip to Content
Merck

Skip To

I8280

Indomethacin

meets USP testing specifications

Synonym(s):

1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H16ClNO4
CAS Number:
Molecular Weight:
357.79
UNSPSC Code:
12352101
NACRES:
NA.21
PubChem Substance ID:
EC Number:
200-186-5
Beilstein/REAXYS Number:
497341
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

USP/NF, meets USP testing specifications

assay

98.0-102.0%

form

powder

mode of action

enzyme | inhibits

SMILES string

COc1ccc2n(c(C)c(CC(O)=O)c2c1)C(=O)c3ccc(Cl)cc3

InChI

1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)

InChI key

CGIGDMFJXJATDK-UHFFFAOYSA-N

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that is relatively selective for COX-1.
Used as a non-selective inhibitor of cyclooxygenases, COX1 and COX2, leading to reduced prostaglandin biosynthesis from arachidonic acid.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I7378405268111902
mode of action

enzyme | inhibits

mode of action

enzyme | inhibits

mode of action

-

mode of action

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

assay

98.0-102.0%

assay

98.0-102.0% (in accordance with EP)

assay

≥98% (by assay)

assay

99%

form

powder

form

powder or crystals

form

powder

form

-

Gene Information

human ... ALB(213), ALOX5(240), ELA2(1991), GPR44(11251), IL1A(3552), PLA2G1B(5319), PTGDR(5729), PTGS1(5742), PTGS2(5743)
mouse ... Alox5(11689), Ptgdr(19214), Ptger1(19216), Ptger2(19217), Ptger3(19218), Ptger4(19219), Ptgs1(19224), Ptgs2(19225)
rat ... Alox5(25290), Ptgs1(24693), Ptgs2(29527)

Gene Information

human ... ALB(213), ALOX5(240), ELA2(1991), GPR44(11251), IL1A(3552), PLA2G1B(5319), PTGDR(5729), PTGS1(5742), PTGS2(5743)
mouse ... Alox5(11689), Ptgdr(19214), Ptger1(19216), Ptger2(19217), Ptger3(19218), Ptger4(19219), Ptgs1(19224)
rat ... Alox5(25290), Ptgs1(24693), Ptgs2(29527)

Gene Information

-

Gene Information

-

agency

USP/NF, meets USP testing specifications

agency

meets EP testing specifications

agency

-

agency

-


Still not finding the right product?

Explore all of our products under Indomethacin


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters