Merck
  • Homing and restorative effects of bone marrow-derived mesenchymal stem cells on cisplatin injured ovaries in rats.
Homing and restorative effects of bone marrow-derived mesenchymal stem cells on cisplatin injured ovaries in rats.
Molecules and cells (2014-11-21)
Jiabin Liu, Haiying Zhang, Yun Zhang, Nan Li, Yuku Wen, Fanglei Cao, Hao Ai, Xiaoou Xue
ABSTRACT
Premature ovarian failure (POF) is a long-term adverse effect of chemotherapy treatment. However, current available treatment regimens are not optimal. Emerging evidence suggests that bone marrow-derived mesenchymal stem cells (BMSCs) could restore the structure and function of injured tissues, but the homing and restorative effects of BMSCs on chemotherapy injured ovaries are still not clear. In this study, we found that granulosa cell (GC) apoptosis induced by cisplatin was reduced when BMSCs were migrated to granulosa cells (GCs) in vitro. Chemotherapy-induced POF was induced by intraperitoneal injection of cisplatin in rats. BMSCs labeled with enhanced green fluorescent protein (EGFP) were injected into the rats via the tail vein to investigate the homing and distribution of BMSCs in vivo. The number of BMSCs in the ovarian hilum and medulla was greater than in the cortex, but no BMSCs were found in the follicles and corpus lutea. In addition, the BMSCs treatment group's antral follicle count and estradiol levels increased after 30 days, compared with the POF group. Hence, our study demonstrates that intravenously delivered BMSCs can home to the ovaries, and restore its structure and function in POF model rats.
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Estradiol, meets USP testing specifications
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
N,N-Dimethylethylenediamine, ≥98.0% (GC)
Sigma-Aldrich
N,N-Dimethylethylenediamine, 95%
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard