Merck
  • Hydrogen-bonded and reduction-responsive micelles loading atorvastatin for therapy of breast cancer metastasis.
Hydrogen-bonded and reduction-responsive micelles loading atorvastatin for therapy of breast cancer metastasis.
Biomaterials (2014-06-06)
Pengfei Xu, Haijun Yu, Zhiwen Zhang, Qingshuo Meng, Huiping Sun, Xianzhi Chen, Qi Yin, Yaping Li
ABSTRACT
Metastasis is one of the major obstacles for the successful therapy of breast cancer. Although increased candidate drugs targeting cancer metastasis are tested, their clinical translation is limited by either serve toxicity or low efficacy. In present work, a nano-drug delivery system loading atorvastatin calcium (Ator) was developed for the efficient suppression of the metastasis of breast cancer. The nano-drug delivery system was constructed by a amphiphilic copolymer of methoxy polyethylene glycol-s-s-vitamin E succinate (mPEG-s-s-VES, PSV), which was consisted of a hydrophilic mPEG1k segment and a hydrophobic VES head, which were conjugated with a linker bearing amide and disulfide groups simultaneously. Self-assembly of PSV and Ator formed Ator-loaded PSV micelles (ASM) with good colloidal stability, high drug loading content (up to 50%) and great encapsulation efficiency (99.09 ± 0.28%). In cellular level, it was found that the ASM could efficiently release the Ator payload into cytosol due to detachment of PEG shell at high intracellular glutathione condition. ASM could significantly inhibit the migration and invasion of 4T1 breast cancer cells with inhibitory rates of 79.2% and 88.5%, respectively. In a 4T1 orthotropic mammary tumor metastatic cancer model, it was demonstrated that ASM could completely blocked the lung and liver metastasis of breast cancer with minimal toxicity owing to enhanced Ator accumulation in tumor and lung as compared with that of free Ator. The down-regulations of metastasis-promoting MMP-9, Twist and uPA proteins were demonstrated as the main underlying mechanism. As a result, ASM could be a promising drug delivery system for the efficient therapy of breast cancer metastasis.
Product Description
Supelco
D-α-Tocopherol succinate, analytical standard
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
D-α-Tocopherol succinate, BioXtra, ≥98.0% (HPLC)
Sigma-Aldrich
D-α-Tocopherol succinate, semisynthetic, 1210 IU/g
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
Tocopheryl Acid Succinate, a, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Hematoxylin