Merck
HSPD1 interacts with IRF3 to facilitate interferon-beta induction.
PloS one (2014-12-17)
Lan Lin, Shan Pan, Jianqing Zhao, Cheng Liu, Pingan Wang, Lei Fu, Xinlin Xu, Meilin Jin, Anding Zhang
ABSTRACT
The production of IFN- I (IFN-α/β) is one of the earliest and most important host-protective responses. Interferon regulatory factor 3 (IRF3) is a critical transcriptional factor in the IFN-β signaling pathway. Although significant progress has been achieved in the regulation of IRF3, the process may be more complicated than previously considered. In the present study, heat shock protein 60 (HSP60, HSPD1) was identified as a novel IRF3-interacting protein. Overexpression of HSPD1 facilitated the phosphorylation and dimerization of IRF3 and enhanced IFN-β induction induced by SeV infection. In contrast, knockdown of endogenous HSPD1 significantly inhibited the signaling pathway. Furthermore, HSPD1 enhanced activation of the IFN-β promoter mediated by RIG-I, MDA-5, MAVS, TBK1 and IKKε but not IRF3/5D, a mock phosphorylated form of IRF3. The present study indicated that HSPD1 interacted with IRF3 and it contributed to the induction of IFN-β.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
SAFC
Glycine
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%