Merck
  • Activation of mesocorticolimbic reward circuits for assessment of relief of ongoing pain: a potential biomarker of efficacy.
Activation of mesocorticolimbic reward circuits for assessment of relief of ongoing pain: a potential biomarker of efficacy.
Pain (2014-05-28)
Jennifer Y Xie, Chaoling Qu, Amol Patwardhan, Michael H Ossipov, Edita Navratilova, Lino Becerra, David Borsook, Frank Porreca
ABSTRACT
Preclinical assessment of pain has increasingly explored operant methods that may allow behavioral assessment of ongoing pain. In animals with incisional injury, peripheral nerve block produces conditioned place preference (CPP) and activates the mesolimbic dopaminergic reward pathway. We hypothesized that activation of this circuit could serve as a neurochemical output measure of relief of ongoing pain. Medications commonly used clinically, including gabapentin and nonsteroidal anti-inflammatory drugs (NSAIDs), were evaluated in models of post-surgical (1 day after incision) or neuropathic (14 days after spinal nerve ligation [SNL]) pain to determine whether the clinical efficacy profile of these drugs in these pain conditions was reflected by extracellular dopamine (DA) release in the nucleus accumbens (NAc) shell. Microdialysis was performed in awake rats. Basal DA levels were not significantly different between experimental groups, and no significant treatment effects were seen in sham-operated animals. Consistent with clinical observation, spinal clonidine produced CPP and produced a dose-related increase in net NAc DA release in SNL rats. Gabapentin, commonly used to treat neuropathic pain, produced increased NAc DA in rats with SNL but not in animals with incisional, injury. In contrast, ketorolac or naproxen produced increased NAc DA in animals with incisional but not neuropathic pain. Increased extracellular NAc DA release was consistent with CPP and was observed selectively with treatments commonly used clinically for post-surgical or neuropathic pain. Evaluation of NAc DA efflux in animal pain models may represent an objective neurochemical assay that may serve as a biomarker of efficacy for novel pain-relieving mechanisms.
Product Description
Clonidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzoylecgonine-d3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Benzoylecgonine-d3 solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Clonidine hydrochloride, solid
Sigma-Aldrich
Naproxen sodium, meets USP testing specifications
Sigma-Aldrich
Naproxen sodium, 98.0-102.0%
Ketorolac trometamol for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Naproxen sodium, United States Pharmacopeia (USP) Reference Standard
USP
Ketorolac Tromethamine, United States Pharmacopeia (USP) Reference Standard
Supelco
Naproxen sodium, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ketorolac Tromethamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ketorolac tris salt, ≥99%, crystalline
Ketorolac trometamol, European Pharmacopoeia (EP) Reference Standard