Merck
  • A novel method for the identification of inorganic and organic gunshot residue particles of lead-free ammunitions from the hands of shooters using scanning laser ablation-ICPMS and Raman micro-spectroscopy.
A novel method for the identification of inorganic and organic gunshot residue particles of lead-free ammunitions from the hands of shooters using scanning laser ablation-ICPMS and Raman micro-spectroscopy.
The Analyst (2014-10-11)
Zuriñe Abrego, Nagore Grijalba, Nora Unceta, Maite Maguregui, Alicia Sanchez, Alberto Fernández-Isla, M Aranzazu Goicolea, Ramón J Barrio
ABSTRACT
A method based on scanning laser ablation and inductively coupled plasma-mass spectrometry (SLA-ICPMS) and Raman micro-spectroscopy for the detection and identification of compounds consistent with gunshot residue particles (GSR) has been developed. The method has been applied to the characterization of particles resulting from the discharge of firearms using lead-free ammunition. Modified tape lifts were used to collect the inorganic and organic residues from skin surfaces in a single sample. Using SLA-ICPMS, aggregates related to the composition of the ammunition, such as Cu-Zn-Sn, Zr-Sr, Cu-Zn, Al-Ti, or Al-Sr-Zr were detected, but this composition is only consistent with GSR from lead-free ammunitions. Additional evidence was provided by micro-Raman spectroscopy, which identified the characteristic organic groups of the particles as centralite, diphenylamine or their nitrated derivatives, which are indicative of GSR.
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Diphenylamine, ACS reagent, ≥99%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Barium nitrate, 99.999% trace metals basis
Supelco
Diphenylamine, PESTANAL®, analytical standard
Sigma-Aldrich
Diphenylamine, ReagentPlus®, 99%
Sigma-Aldrich
Diphenylamine, puriss. p.a., redox indicator, ACS reagent, reag. Ph. Eur., ≥98% (GC)
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Barium nitrate, ACS reagent, ≥99%
Sigma-Aldrich
Diethyl phthalate, ≥99%
Sigma-Aldrich
4-Nitrodiphenylamine, 99%
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Nitroguanidine
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Diethyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Diethyl Phthalate, Pharmaceutical Secondary Standard; Certified Reference Material