Merck
  • Embryological, clinical and ultrastructural study of human oocytes presenting indented zona pellucida.
Embryological, clinical and ultrastructural study of human oocytes presenting indented zona pellucida.
Zygote (Cambridge, England) (2013-09-03)
M Sousa, J Teixeira da Silva, J Silva, M Cunha, P Viana, E Oliveira, R Sá, C Soares, C Oliveira, A Barros
ABSTRACT
Human oocyte dysmorphisms attain a large proportion of retrieved oocytes from assisted reproductive technology (ART) treatment cycles. Extracytoplasmic defects involve abnormal morphology of the zona pellucida (ZP), perivitelline space and first polar body. The aim of the present study was to describe a novel dysmorphism affecting the ZP, indented ZP. We also evaluated the clinical, embryological and ultrastructural features of these cases. We evaluated all ART treatment cycles during 7 consecutive years and found 13 treatment cycles (six patients) with all oocytes presenting an indented ZP. In addition, these oocytes presented total or partial absence of the perivitelline space, absence of resistance to ZP and oolemma penetration during microinjection, and low ooplasm viscosity during aspiration. This novel described dysmorphism was recurrent and attained all oocytes in three cases that had more than one treatment cycle. When compared with controls, data showed significant low oocyte maturity (42% versus 81.6%) and high cycle cancellation (30.8% versus 8.5%) rates, normal degeneration (3.4% versus 6.3%) and fertilization rates (69% versus 69.5%), and low pregnancy (15.4% versus 33.3%) and live-birth delivery (7.7% versus 27.7%) rates per cycle. Ultrastructure analysis revealed a zona pellucida structure with large empty electrolucent regions, an outer ZP layer with an indented surface with protuberances and a thick inner ZP that obliterated the perivitelline space. There was evidence of exocytosis of ZP material by the oocyte. In conclusion, oocytes with this novel described dysmorphism (indented ZP) are associated with low maturity, pregnancy and live-birth delivery rates.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Luteinizing hormone releasing hormone, ≥97% (HPLC)
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Estradiol hemihydrate, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications