Merck
  • Volatile profiles of members of the USDA Geneva Malus Core Collection: utility in evaluation of a hypothesized biosynthetic pathway for esters derived from 2-methylbutanoate and 2-methylbutan-1-ol.
Volatile profiles of members of the USDA Geneva Malus Core Collection: utility in evaluation of a hypothesized biosynthetic pathway for esters derived from 2-methylbutanoate and 2-methylbutan-1-ol.
Journal of agricultural and food chemistry (2015-02-05)
Nobuko Sugimoto, Philip Forsline, Randolph Beaudry
ABSTRACT
The volatile ester and alcohol profiles of ripening apple fruit from 184 germplasm lines in the USDA Malus Germplasm Repository at the New York Agricultural Experiment Station in Geneva, NY, USA, were evaluated. Cluster analysis suggested biochemical relationships exist between several ester classes. A strong linkage was revealed between 2-methylbutanoate, propanoate, and butanoate esters, suggesting the influence of the recently proposed "citramalic acid pathway" in apple fruit. Those lines with a high content of esters formed from 2-methylbutan-1-ol and 2-methylbutanoate (2MB) relative to straight-chain (SC) esters (high 2MB/SC ratio) exhibited a marked increase in isoleucine and citramalic acid during ripening, but those lines with a low content did not. Thus, the data were consistent with the existence of the hypothesized citramalic acid pathway and suggest that the Geneva Malus Germplasm Repository, appropriately used, could be helpful in expanding our understanding of mechanisms for fruit volatile synthesis and other aspects of secondary metabolism.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
(±)-2-Methyl-1-butanol, ≥98.0% (GC)
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
SAFC
L-Isoleucine
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
DL-Valine, ≥97%
Sigma-Aldrich
DL-Valine, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)