Merck
  • New molecular staging with G-factor supplements TNM classification in gastric cancer: a multicenter collaborative research by the Japan Society for Gastroenterological Carcinogenesis G-Project committee.
New molecular staging with G-factor supplements TNM classification in gastric cancer: a multicenter collaborative research by the Japan Society for Gastroenterological Carcinogenesis G-Project committee.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association (2014-02-04)
Tetsuji Sawada, Masakazu Yashiro, Kazuhiro Sentani, Naohide Oue, Wataru Yasui, Kohji Miyazaki, Keita Kai, Sachio Fushida, Takashi Fujimura, Masaichi Ohira, Yoshihiro Kakeji, Shoji Natsugoe, Ken Shirabe, Sachiyo Nomura, Yutaka Shimada, Naohiro Tomita, Kosei Hirakawa, Yoshihiko Maehara
ABSTRACT
The G-Project committee was erected by the Japan Society for Gastroenterological Carcinogenesis with an aim of establishing a new classification scheme based on molecular biological characteristics that would supplement the conventional TNM classification to better predict outcome. In a literature search involving 822 articles on gastric cancer, eight molecules including p53, vascular endothelial growth factor (VEGF)-A, VEGF-C, matrix metalloproteinase-7 (MMP-7), human epidermal growth factor receptor 2, Regenerating islet-derived family, member 4, olfactomedin-4 and Claudin-18 were selected as candidates to be included in the new molecular classification scheme named G-factor. A total of 210 cases of gastric cancer who underwent curative R0 resection were registered from four independent facilities. Immunohistochemical staining for the aforementioned molecules was performed for the surgically resected specimens of the 210 cases to investigate the correlation between clinicopathological factors and expression of each molecule. No significant correlation was observed between the immunostaining expression of any of the eight factors and postoperative recurrence. However, the expressions of p53 and MMP-7 were significantly correlated with overall survival (OS). When 210 gastric cancer patients were divided into three groups based on the expression of p53 and MMP-7 (G0 group: negative for both p53 and MMP-7, n = 69, G1 group: positive for either p53 or MMP-7, n = 97, G2 group: positive for both of the molecules, n = 44), G2 group demonstrated significantly higher recurrence rate (59%) compared to 38% in G0 (p = 0.047). The multivariate regression analysis revealed that G2 group was independently associated with a shorter disease-free survival (DFS) (hazard ratio 1.904, 95% CI 1.098-3.303; p = 0.022), although the association with OS was not significant. Stage II patients among the G2 group had significantly inferior prognosis both in terms of OS and DFS when compared with those among the G0/G1 group, with survival curves similar to those of Stage III cases. G-factor based on the expression of p53 and MMP-7 was found to be a promising factor to predict outcome of Stage II/III gastric cancer, and possibly to help select the treatment for Stage II cancer, thus supplementing the conventional TNM system.
Product Description
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)