Merck
  • Guiding principles for site selective and stereoselective intermolecular C-H functionalization by donor/acceptor rhodium carbenes.
Guiding principles for site selective and stereoselective intermolecular C-H functionalization by donor/acceptor rhodium carbenes.
Chemical Society reviews (2011-03-02)
Huw M L Davies, Daniel Morton
ABSTRACT
This tutorial review presents a description of the controlling elements of intermolecular C-H functionalization by means of C-H insertion by donor/acceptor rhodium carbenes. These rhodium carbenes, readily derived from the combination of diazo compounds with dirhodium(ii) catalysts, are sufficiently reactive to undergo a wide range of C-H insertions. They are also capable of highly selective reactions, controlled by a combination of steric and electronic factors. An overview of the structural factors that influence site selectivity will be given, followed by a description of the exceptional diastereo- and enantioselectivity that can be achieved. Several examples will be shown of how this methodology can be applied to streamline the synthesis of natural products and pharmaceutical targets.
Product Description
Rhodium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%
Rhodium, wire reel, 0.1m, diameter 0.25mm, hard, 99.9%
Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.5m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.2m, diameter 0.12mm, as drawn, 99.9%
Rhodium, wire reel, 0.2m, diameter 0.25mm, hard, 99.9%
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis
Rhodium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.85%
Rhodium, microfoil, disks, 10mm, thinness 0.50μm, specific density 620μg/cm2, permanent mylar 3.5μm support, 99.99%
Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.006mm, as rolled, 99.9%
Rhodium, microfoil, disks, 25mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, x-ray quality, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.25mm, hard, 99.9%
Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Rhodium, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.9%
Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.85%
Rhodium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rhodium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%
Rhodium, foil, not light tested, 50x50mm, thickness 0.006mm, as rolled, 99.9%
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rhodium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9%