Skip to Content
Merck
All Photos(2)

Key Documents

GF17496973

Rhodium

wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%

Synonym(s):

Rhodium, RH005120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Rh
CAS Number:
Molecular Weight:
102.91
MDL number:
UNSPSC Code:
12141738
PubChem Substance ID:
NACRES:
NA.23

Assay

99.90%

form

wire

manufacturer/tradename

Goodfellow 174-969-73

resistivity

4.33 μΩ-cm, 20°C

L × diam.

0.2 m × 0.5 mm

bp

3727 °C (lit.)

mp

1966 °C (lit.)

density

12.41 g/cm3 (lit.)

SMILES string

[Rh]

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huw M L Davies et al.
Chemical Society reviews, 36(7), 1109-1119 (2007-06-20)
This tutorial review describes the reactions of the electron-rich heterocycles pyrrole, furan, indole and benzofuran with copper and rhodium carbenoids. Two main reaction pathways are possible, involving either a concerted non-synchronous cyclopropanation or zwitterionic intermediates. A diverse range of products
Jean Bouffard et al.
Topics in current chemistry, 292, 231-280 (2010-01-01)
A review is presented of synthetic methods for the preparation of biaryls by the rhodium-catalyzed C-H bond arylation of arenes with aryl halides (C-H/ C-X couplings), arylmetal reagents (C-H/C-M couplings) and arenes (C-H/C-H couplings), with an emphasis on postulated mechanisms
Albert Padwa
Chemical Society reviews, 38(11), 3072-3081 (2009-10-23)
In this tutorial review, the rhodium(II)-catalyzed domino reactions of alpha-diazo carbonyl compounds are summarized. The article will emphasize some of the more recent synthetic applications of the rhodium carbenoid cyclization/cycloaddition domino cascade for alkaloid synthesis. The many structurally diverse and
Rhodium-catalyzed C-C bond formation via heteroatom-directed C-H bond activation.
Denise A Colby et al.
Chemical reviews, 110(2), 624-655 (2009-05-15)
Jerzy Klosin et al.
Accounts of chemical research, 40(12), 1251-1259 (2007-11-14)
A series of bis-phosphite and bis-phosphine ligands for asymmetric hydroformylation reactions has been evaluated. Bis-phosphite ligands lead, in general, to high regioselectivities across a range of substrates while good enantioselectivities are limited to only a few examples. We found that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service