Merck
  • Biologic properties of eugenol and zinc oxide-eugenol. A clinically oriented review.
Biologic properties of eugenol and zinc oxide-eugenol. A clinically oriented review.
Oral surgery, oral medicine, and oral pathology (1992-06-01)
K Markowitz, M Moynihan, M Liu, S Kim
ABSTRACT
Eugenol-containing dental materials are frequently used in clinical dentistry. When zinc oxide-eugenol (ZOE) is applied to a dentinal cavity, small quantities of eugenol diffuse through the dentin to the pulp. Low concentrations of eugenol exert anti-inflammatory and local anesthetic effects on the dental pulp. Thus use of ZOE temporary filling may facilitate pulpal healing; on the other hand, high eugenol concentrations are cytotoxic. Direct application of eugenol to pulp tissue may result in extensive tissue damage. The ability of ZOE-based endodontic sealers to influence periapical tissue healing is considered in view of eugenol's anti-inflammatory and toxic properties.
Product Description
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Eugenol, European Pharmacopoeia (EP) Reference Standard