Merck
  • Transcriptional activation of genes by 17 beta-estradiol through estrogen receptor-Sp1 interactions.
Transcriptional activation of genes by 17 beta-estradiol through estrogen receptor-Sp1 interactions.
Vitamins and hormones (2001-05-11)
S Safe
ABSTRACT
Estrogen receptor-alpha (ER alpha) is a ligand-activated transcription factor and a member of the nuclear receptor superfamily. The classic mechanism of ER alpha action is associated with estrogen-induced formation of a nuclear ER alpha homodimer, binding to 5'-regulatory estrogen response elements (EREs) in target gene promoters, interaction with other nuclear proteins, and general transcription factors to activate gene expression. ER alpha also interacts with Sp1 protein to transactivate genes through binding Sp1(N)xERE or Sp1(N)xERE half-site (1/2) motifs where both ER alpha and Sp1 bind DNA elements. Activation through Sp1(N)xERE1/2 requires interactions of both proteins with their cognate DNA elements as well as additional nuclear factors to form a functional ER alpha/Sp1-DNA complex. Recent studies also show that ER alpha and Sp1 physically interact and ER alpha preferentially binds to the C-terminal DNA-binding domain of Sp1 protein. Moreover, ER alpha/Sp1 can activate transcription from a consensus GC-rich Sp1 binding site in transient transfection studies in MCF-7 human breast cancer cells, and this response is also observed with ER alpha variants that do not contain the DNA-binding domain. Several genes that are induced by estrogens in MCF-7 cells are activated through one or more GC-rich sites in their regulatory regions and these include the cathepsin D, E2F1, bcl-2, c-fos, adenosine deaminase, insulinlike growth factor binding protein 4, and retinoic acid receptor alpha 1 genes. ER alpha/Sp1 and ER beta/Sp1 action is dependent on ligand structure and cell context and ER beta/Sp1 is primarily associated with decreased ligand-dependent gene expression. ER alpha/Sp1, like ER alpha/AP1, represents a pathway for hormone activation of genes in which the receptor does not bind DNA, and results of ongoing studies suggest that ER alpha/Sp1 plays an important role in transcriptional activation of multiple growth regulatory genes in breast cancer cells.
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material