Merck
  • Homopiperazine grafted mesoporous silicas from rice husk ash for CO2 adsorption.
Homopiperazine grafted mesoporous silicas from rice husk ash for CO2 adsorption.
Journal of nanoscience and nanotechnology (2014-04-18)
Rajangam Vinodh, Margandan Bhagiyalakshmi, Pushparaj Hemalatha, Mani Ganesh, Mei Mei Peng, Muthiahpillai Palanichamy, Wang Seog Cha, Hyun Tae Jang
ABSTRACT
Chloro-functionalized mesoporous MCM-41, SBA-15, MCM-48 and KIT-6 were synthesized by co-condensation of 3-chloropropyl-trimethoxy-silane (CPTMS) and rice husk ash sodium silicate solution, which is subsequently grafted with a heterocyclic amine, homopiperazine (HPZ). X-ray powder diffraction and BET analysis of the chloro-functionalized mesoporous silicas confirmed the similarity between their structural properties and those obtained from conventional silica sources. CO2 adsorption studies of all HPZ-grafted mesoporous silicas exhibited 8-10 wt% of adsorption capacity and are found to be selective, recyclable and thermally stable. Here, the CO2 adsorption reaction is via the traditional carbamate mechanism. The presence of both secondary and tertiary amine in HPZ influences the high CO2 adsorption capacity. Hence, these HPZ-grafted mesoporous silicas could contribute to CO2 capture as a green, tunable, selective and efficient sorbent.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Supelco
Piperazine hexahydrate, analytical standard
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Piperazine hydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O