Merck
  • Role of solution pH on the microstructural properties of spin coated cobalt oxide thin films.
Role of solution pH on the microstructural properties of spin coated cobalt oxide thin films.
Journal of nanoscience and nanotechnology (2014-04-18)
S Valanarasu, V Dhanasekaran, M Karunakaran, R Chandramohan, T Mahalingam
ABSTRACT
Cobalt Oxide (Co3o4) thin films have been successfully coated onto glass substrates at various solution pH by sol-gel spin coating technique. The film thickness was estimated using weight gain method and it revealed that the film thickness increased with solution pH values. The prepared film structural, morphological, optical and electrical properties were studied using X-ray diffraction (XRD), scanning electron microscope (SEM), UV-Vis-NIR spectrophotometer and Vander Pau method, respectively. The structure of the films were found to be face centered cubic with preferential orientation along (311) plane. X-ray line profile analysis was used to evaluate the micro structural parameters such as crystallite size, micro strain, dislocation density and stacking fault probability. The crystallite size values are increased with increase of solution pH values and maximum value of crystallite is estimated at 40.8 nm at solution pH 8 +/- 0.1. Morphological results showed that the pH of the solution has a marked effect on morphology of the Co3O4 thin films. The optical studies revealed that the band gap can be tailored between 2.16 to 2.31 eV by altering pH. The thin film formed at a solution pH 7 is found to have a low resistivity and high mobility. The electrical resistivity (p), carrier concentration (n) and mobility (micro) values are 0.1 x 10(3) omega x cm, 8.9 cm2 gammas(-1) and 6.6 x 10(14) cm(-3), respectively for Co3O4 thin film prepared at solution pH 7 +/- 0.1. EDAX studies showed that the cobalt content increased and the oxygen content decreased with increase of pH.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Cobalt, foil, light tested, 50x50mm, thickness 0.009mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.008mm, 99.9%
Cobalt, foil, not light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 150x150mm, thickness 0.015mm, 99.9%
Cobalt, foil, 10mm disks, thickness 0.008mm, 99.9%
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Cobalt, foil, 50x50mm, thickness 0.5mm, as rolled, 99.99+%
Cobalt, foil, 25mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Cobalt, foil, 15mm disks, thickness 0.008mm, 99.9%
Cobalt, foil, 10mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, 10mm disks, thickness 0.02mm, 99.9%
Cobalt, foil, 15mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, 50x50mm, thickness 0.7mm, annealed, 99.9%
Cobalt, foil, 100x100mm, thickness 1.0mm, as rolled, 99.9%
Cobalt, foil, 25mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Cobalt, foil, 15mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 15mm disks, thickness 0.009mm, 99.9%