Merck
  • Measurement of histidine pKa values and tautomer populations in invisible protein states.
Measurement of histidine pKa values and tautomer populations in invisible protein states.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-16)
Alexandar L Hansen, Lewis E Kay
ABSTRACT
The histidine imidazole side chain plays a critical role in protein function and stability. Its importance for catalysis is underscored by the fact that histidines are localized to active sites in ∼ 50% of all enzymes. NMR spectroscopy has become an important tool for studies of histidine side chains through the measurement of site-specific pK(a)s and tautomer populations. To date, such studies have been confined to observable protein ground states; however, a complete understanding of the role of histidine electrostatics in protein function and stability requires that similar investigations be extended to rare, transiently formed conformers that populate the energy landscape, yet are often "invisible" in standard NMR spectra. Here we present NMR experiments and a simple strategy for studies of such conformationally excited states based on measurement of histidine (13)Cγ, (13)Cδ2 chemical shifts and (1)Hε-(13)Cε one-bond scalar couplings. The methodology is first validated and then used to obtain pKa values and tautomer distributions for histidine residues of an invisible on-pathway folding intermediate of the colicin E7 immunity protein. Our results imply that the side chains of H40 and H47 are exposed in the intermediate state and undergo significant conformational rearrangements during folding to the native structure. Further, the pKa values explain the pH-dependent stability differences between native and intermediate states over the pH range 5.5-6.5 and they suggest that imidazole deprotonation is not a barrier to the folding of this protein.
Product Description
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Imidazole, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Histidine
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Histidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O