Merck
  • Protein-enriched diet, with the use of lean red meat, combined with progressive resistance training enhances lean tissue mass and muscle strength and reduces circulating IL-6 concentrations in elderly women: a cluster randomized controlled trial.
Protein-enriched diet, with the use of lean red meat, combined with progressive resistance training enhances lean tissue mass and muscle strength and reduces circulating IL-6 concentrations in elderly women: a cluster randomized controlled trial.
The American journal of clinical nutrition (2014-01-31)
Robin M Daly, Stella L O'Connell, Niamh L Mundell, Carley A Grimes, David W Dunstan, Caryl A Nowson
ABSTRACT
Physical inactivity, inadequate dietary protein, and low-grade systemic inflammation contribute to age-related muscle loss, impaired function, and disability. We assessed the effects of progressive resistance training (PRT) combined with a protein-enriched diet facilitated through lean red meat on lean tissue mass (LTM), muscle size, strength and function, circulating inflammatory markers, blood pressure, and lipids in elderly women. In a 4-mo cluster randomized controlled trial, 100 women aged 60-90 y who were residing in 15 retirement villages were allocated to receive PRT with lean red meat (∼160 g cooked) to be consumed 6 d/wk [resistance training plus lean red meat (RT+Meat) group; n = 53] or control PRT [1 serving pasta or rice/d; control resistance training (CRT) group; n = 47)]. All women undertook PRT 2 times/wk and received 1000 IU vitamin D3/d. The mean (± SD) protein intake was greater in the RT+Meat group than in the CRT group throughout the study (1.3 ± 0.3 compared with 1.1 ± 0.3 g · kg⁻¹ · d⁻¹, respectively; P < 0.05). The RT+Meat group experienced greater gains in total body LTM (0.45 kg; 95% CI: 0.07, 0.84 kg), leg LTM (0.22 kg; 95% CI: 0.02, 0.42 kg), and muscle strength (18%; 95% CI: 0.03, 0.34) than did the CRT group (all P < 0.05). The RT+Meat group also experienced a 10% greater increase in serum insulin-like growth factor I (P < 0.05) and a 16% greater reduction in the proinflammatory marker interleukin-6 (IL-6) (P < 0.05) after 4 mo. There were no between-group differences for the change in blood lipids or blood pressure. A protein-enriched diet equivalent to ∼1.3 g · kg⁻¹ · d⁻¹ achieved through lean red meat is safe and effective for enhancing the effects of PRT on LTM and muscle strength and reducing circulating IL-6 concentrations in elderly women. This trial was registered at the Australian Clinical Trials Registry as ACTRN12609000223235.
Product Description
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, analytical standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®