Skip to Content
Merck
All Photos(1)

Key Documents

C9774

Sigma-Aldrich

Cholecalciferol

analytical standard

Synonym(s):

(+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Beilstein:
2339331
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058003
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic - wool)

Quality Level

grade

analytical standard

Assay

≥97.0%

form

crystalline

packaging

pkg of 250 mg

color

white

mp

83-86 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26+,27-/m1/s1

InChI key

QYSXJUFSXHHAJI-YRZJJWOYSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

Cholecalciferol, also called vitamin D3, promotes the absorption of calcium and phosphorus in the body to regulate bone growth. It is most commonly found in humans and animals and is produced in our skin when exposed to sunlight. It is used to treat and prevent bone disorders (such as rickets, osteomalacia).

Application

Cholecalciferol has been used as an internal standard in high-performance liquid chromatography (HPLC) to analyze serum vitamin D levels. It has also been used as a standard in liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) to quantify vitamin D3 in sunflower oil.
Cholecalciferol may be used as an analytical reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography.

Biochem/physiol Actions

Cholecalciferol is sourced either through the diet or exposing skin to ultraviolet rays. Oral administration of vitamin D3 is readily absorbed and is stored in adipose tissue. Deficiency of vitamin D is often observed in chronic kidney disease.
Vitamin D acts through a receptor that is a member of the ligand-dependent transcription factor superfamily. Modulates the proliferation and differentiation of both normal and cancer cells. Has antiproliferative and antimetastatic effects on breast, colon, and prostate cancer cells. Activated vitamin D receptors in intestine and bone maintain calcium absorbance and homeostasis.
Vitamin D acts through a receptor that modulates the proliferation and differentiation of both normal and cancer cells.

Packaging

Packaged in flame-sealed ampules under Argon

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability of vitamin D3 and vitamin D2 in oil, fish and mushrooms after household cooking
Lovznjak P and Jakobsen J
Food Chemistry, 254(4), 144-149 (2018)
Vitamin D serum levels are associated with handgrip strength but not with muscle mass or length of hospital stay after hip fracture
Gumieiro DN, et al.
Nutrition, 31(7-8), 931-934 (2015)
Genetic polymorphisms of vitamin D metabolism genes and serum level of vitamin D in colorectal cancer
Vidigal VM, et al.
The International Journal of Biological Markers, 32(4), 441-446 (2017)
Clinical Veterinary Toxicology, 448-448 (2003)
Cholecalciferol supplementation in hemodialysis patients: effects on mineral metabolism, inflammation, and cardiac dimension parameters
Matias P, et al.
Clinical journal of the American Society of Nephrology : CJASN (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service