Merck
  • Antioxidant supplements reduced oxidative stress and stabilized liver function tests but did not reduce inflammation in a randomized controlled trial in obese children and adolescents.
Antioxidant supplements reduced oxidative stress and stabilized liver function tests but did not reduce inflammation in a randomized controlled trial in obese children and adolescents.
The Journal of nutrition (2013-12-20)
Stefanie B Murer, Isabelle Aeberli, Christian P Braegger, Matthias Gittermann, Martin Hersberger, Scott W Leonard, Alan W Taylor, Maret G Traber, Michael B Zimmermann
ABSTRACT
Oxidative stress and low-grade systemic inflammation may contribute to the pathogenesis of obesity-induced comorbidities, including nonalcoholic fatty liver disease. Increasing intake of dietary antioxidants might be beneficial, but there are few data in obese children. To examine the effect of antioxidant supplementation on biomarkers of oxidative stress, inflammation, and liver function, we randomly assigned overweight or obese children and adolescents (n = 44; mean ± SD age: 12.7 ± 1.5 y) participating in a lifestyle modification program to a 4-mo intervention with daily antioxidants (vitamin E, 400 IU; vitamin C, 500 mg; selenium, 50 μg) or placebo. We measured anthropometrics, antioxidant status, oxidative stress (F(2)-isoprostanes, F(2)-isoprostane metabolites), inflammation, liver enzymes, fasting insulin and glucose, and lipid profile at baseline and endpoint. There was a significant treatment effect of antioxidant supplementation on antioxidant status [α-tocopherol, β = 23.2 (95% CI: 18.0, 28.4); ascorbic acid, β = 70.6 (95% CI: 51.7, 89.4); selenium, β = 0.07 (95% CI: 0.01, 0.12)] and oxidative stress [8-iso-prostaglandin F2α, β = -0.11 (95% CI: -0.19, -0.02)] but not on any of the inflammatory markers measured. There was a significant treatment effect on alanine aminotransferase [β = -0.13 (95% CI: -0.23, -0.03)], a trend toward a significant effect on aspartate aminotransferase [β = -0.04 (95% CI: -0.09, 0.01)], and no significant effect on γ-glutamyltransferase [β = -0.03 (95% CI: -0.11, 0.06)]. In summary, antioxidant supplementation for 4 mo improved antioxidant-oxidant balance and modestly improved liver function tests; however, it did not reduce markers of systemic inflammation despite significant baseline correlations between oxidative stress and inflammation. The study was registered at clinicaltrials.gov as NCT01316081.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
γ-Glutamyl Transferase, ERM®, certified reference material
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland