Merck
  • Effect of cellulose reinforcement on the properties of organic acid modified starch microparticles/plasticized starch bio-composite films.
Effect of cellulose reinforcement on the properties of organic acid modified starch microparticles/plasticized starch bio-composite films.
Carbohydrate polymers (2013-03-08)
Carmen-Alice Teacă, Ruxanda Bodîrlău, Iuliana Spiridon
ABSTRACT
The present paper describes the preparation and characterization of polysaccharides-based bio-composite films obtained by the incorporation of 10, 20 and 30 wt% birch cellulose (BC) within a glycerol plasticized matrix constituted by the corn starch (S) and chemical modified starch microparticles (MS). The obtained materials (coded as MS/S, respectively MS/S/BC) were further characterized. FTIR spectroscopy and X-ray diffraction were used to evidence structural and crystallinity changes in starch based films. Morphological, thermal, mechanical, and water resistance properties were also investigated. Addition of cellulose alongside modified starch microparticles determined a slightly improvement of the starch-based films water resistance. Some reduction of water uptake for any given time was observed mainly for samples containing 30% BC. Some compatibility occurred between MS and BC fillers, as evidenced by mechanical properties. Tensile strength increased from 5.9 to 15.1 MPa when BC content varied from 0 to 30%, while elongation at break decreased significantly.
Sigma-Aldrich
DL-Tartaric acid, ≥99%
Sigma-Aldrich
DL-Tartaric acid, ReagentPlus®, 99%
Sigma-Aldrich
D-(−)-Tartaric acid, puriss., unnatural form, ≥99.0% (T)
Sigma-Aldrich
L-(+)-Tartaric acid, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
D-(−)-Tartaric acid, ReagentPlus®, 99%
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.7%, FCC, FG
Sigma-Aldrich
L-(+)-Tartaric acid, BioXtra
Supelco
L-(+)-Tartaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-(+)-Tartaric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%