Skip to Content
Merck
All Photos(4)

Key Documents

798665

Sigma-Aldrich

L-(+)-Tartaric acid

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous

Quality Level

vapor density

5.18 (vs air)

product line

Redi-Dri

Assay

≥99.5%

form

crystalline powder
crystals

optical activity

[α]20/D +12.4°, c = 20 in H2O

optical purity

ee: 99% (GLC)

autoignition temp.

797 °F

quality

free-flowing

impurities

≤0.002% S compounds
≤0.005% insolubles

ign. residue

≤0.02%

mp

170-172 °C (lit.)

solubility

water: soluble 150 g/L at 25 °C

anion traces

chloride (Cl-): ≤0.001%
oxalate (C2O42-): passes test
phosphate (PO43-): ≤0.001%

cation traces

Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral building block for natural products. Also forms a Diels-Alder catalyst with TiCl2(O-i-Pr)2.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry Letters (Jpn), 1967-1967 (1986)
Chemistry Letters (Jpn), 2409-2409 (1987)
Organic acid profile of Turkish white grapes and grape juices.
Soyer Y, et al.
J. Food Compos. Anal., 16(5), 629-636 (2003)
Asymmetric synthesis of both enantiomers of pyrrolidinoisoquinoline derivatives from L-malic acid and L-tartaric acid.
Lee YS, et al.
The Journal of Organic Chemistry, 60(22), 7149-7152 (1955)
Organic salts of L-tartaric acid: materials for second harmonic generation with a crystal structure governed by an anionic hydrogen-bonded network.
Aakeroy CB, et al.
Journal of the Chemical Society. Chemical Communications, 7, 553-555 (1992)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service