Merck
  • Fully automated synthesis of 4-[18F]fluorobenzylamine based on borohydride/NiCl2 reduction.
Fully automated synthesis of 4-[18F]fluorobenzylamine based on borohydride/NiCl2 reduction.
Nuclear medicine and biology (2013-01-12)
Jenilee Way, Frank Wuest
ABSTRACT
4-[(18)F]Fluorobenzylamine ([(18)F]FBA) is an important building block for the synthesis of (18)F-labeled compounds. Synthesis of [(18)F]FBA usually involves application of strong reducing agents like LiAlH4 which is challenging to handle in automated synthesis units (ASUs). Therefore, alternative methods for the preparation of [(18)F]FBA compatible with remotely-controlled syntheses in ASUs are needed. (18)F]FBA was prepared in a remotely-controlled synthesis unit (GE TRACERlab™ FX) based on Ni(II)-mediated borohydride exchange resin (BER) reduction of 4-[(18)F]fluorobenzonitrile ([(18)F]FBN). [(18)F]FBA was used for the synthesis of novel thiol-reactive prosthetic group 4-[(18)F]fluorobenzyl)maleimide [(18)F]FBM and Hsp90 inhibitor 17-(4-[(18)F]fluorobenzylamino)-17-demethoxy-geldanamycin [(18)F] GA. [(18)F]FBA could be prepared in high radiochemical yield greater than 80% (decay-corrected) within 60min. In a typical experiment, 7.4GBq of [(18)F]FBA could be obtained in high radiochemical purity of greater than 95% starting from 10GBq of cyclotron-produced n.c.a. [(18)F]fluoride. [(18)F]FBA was used for the preparation of 4-[(18)F]fluorobenzyl)maleimide as a novel prosthetic group for labeling of thiol groups as demonstrated with tripeptide glutathione. [(18)F]FBA was also used as building block for the syntheses of small molecules as exemplified by the preparation of Hsp90 inhibitor 17-(4-[(18)F]fluorobenzylamino)-17-demethoxy-geldanamycin. The described remotely-controlled synthesis of [(18)F]FBA will significantly improve the availability of [(18)F]FBA as an important and versatile building block for the development of novel (18)F-labeled compounds containing a fluorobenzylamine moiety.
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.9% trace metals basis
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Nickel(II) chloride hydrate, 99.95% trace metals basis
Sigma-Aldrich
4-Fluorobenzylamine, 97%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Nickel(II) chloride hexahydrate, puriss. p.a., ≥98%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, ReagentPlus®
Sigma-Aldrich
Nickel(II) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Nickel(II) chloride, 98%