Merck
  • [Hydrocortisone 17-butyrate (Locoid), a thirty-year ongoing innovative drug].
[Hydrocortisone 17-butyrate (Locoid), a thirty-year ongoing innovative drug].
Revue medicale de Liege (2006-03-29)
G E Piérard
ABSTRACT
Since half a century, dermocorticoids represent worthy compounds in dermatological therapy. About thirty years ago, hydrocortisone 17-butyrate was introduced on the Belgian market. This compound combines good efficiency and reduced risk of side effects. Its topical formulations have been enriched over time. The original Locoid lipocream and Locoid Crelo presentations confer remarkable qualities to the product. We report a synopsis of clinical and translational studies devoted to this unique dermocorticoid.
Product Description
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone 17-butyrate
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
USP
Hydrocortisone butyrate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard