Merck
Metal loaded zeolite adsorbents for hydrogen cyanide removal.
Journal of environmental sciences (China) (2013-08-09)
Ping Ning, Juan Qiu, Xueqian Wang, Wei Liu, Wei Chen
ABSTRACT
Metal (Cu, Co, or Zn) loaded ZSM-5 and Y zeolite adsorbents were prepared for the adsorption of hydrogen cyanide (HCN) toxic gas. The results showed that the HCN breakthrough capacity was enhanced significantly when zeolites were loaded with Cu. The physical and chemical properties of the adsorbents that influence the HCN adsorption capacity were analyzed. The maximal HCN breakthrough capacities were about the same for both zeolites at 2.2 mol of HCN/mol of Cu. The Cu2p XPS spectra showed that the possible species present were Cu2O and CuO. The N1s XPS data and FT-IR spectra indicated that CN(-) would be formed in the presence of Cu+/Cu2+ and oxygen gas, and the reaction product could be adsorbed onto Cu/ZSM-5 zeolite more easily than HCN.
Product Description
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Copper (O), BCR®, certified reference material, rod
Copper (S, P), BCR®, certified reference material, chips
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Copper, IRMM®, certified reference material, 0.5 mm wire
Copper, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Aluminosilicate, mesostructured, MCM-41 (hexagonal)
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Zeolite
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis