Skip to Content
Merck
All Photos(2)

Key Documents

311405

Sigma-Aldrich

Copper

ACS reagent, granular, 10-40 mesh, ≥99.90%

Synonym(s):

Cu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
EC Number:
MDL number:
UNSPSC Code:
12141711
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥99.90%

form

granular

reaction suitability

core: copper
reagent type: catalyst

resistivity

1.673 μΩ-cm, 20°C

impurities

≤0.001% P
≤0.02% insol. dil. HNO3

particle size

10-40 mesh

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

cation traces

Ag: ≤0.002%
As: ≤5 ppm
Fe: ≤0.005%
Mn: ≤0.001%
Pb: ≤0.005%
Sb and Sn: ≤0.01%

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

Application

Copper (granular) can be used as a catalyst in the hydrogenation of ethylene.[1] It can be impregnated on activated carbon material and used as an adsorbent and also as a catalyst for the removal of SO2 from flue gases.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of occluded hydrogen on the hydrogenation of ethylene over copper
M. C. Schachter, et al.
The Journal of Physical Chemistry, 79(16), 1698?1702-1698?1702 (1975)
Carbon materials as catalyst supports for SO2 oxidation: catalytic activity of CuO-AC
Tseng H-H, et al.
Carbon, 41(1), 139-149 (2003)
Daniel L Priebbenow et al.
Organic letters, 15(24), 6155-6157 (2013-11-28)
A method has been developed for the preparation of N-alkynylated sulfoximines involving the copper-catalyzed decarboxylative coupling of sulfoximines with aryl propiolic acids. A range of substituents on both the sulfoximidoyl moiety and the aryl group of the propiolic acid were
Hiroshi Sato et al.
Science (New York, N.Y.), 343(6167), 167-170 (2013-12-18)
Carbon monoxide (CO) produced in many large-scale industrial oxidation processes is difficult to separate from nitrogen (N2), and afterward, CO is further oxidized to carbon dioxide. Here, we report a soft nanoporous crystalline material that selectively adsorbs CO with adaptable
Magnus Andersson et al.
Nature structural & molecular biology, 21(1), 43-48 (2013-12-10)
Heavy metals in cells are typically regulated by PIB-type ATPases. The first structure of the class, a Cu(+)-ATPase from Legionella pneumophila (LpCopA), outlined a copper transport pathway across the membrane, which was inferred to be occluded. Here we show by

Questions

  1. Is there a provision of activated copper products?

    1 answer
    1. Activated copper products are not included in the portfolio.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service