Merck
  • The content of conjugated linoleic acid (CLA) isomer groups in milk of two Polish sheep breeds determined by silver ion liquid chromatography (Ag(+)-HPLC).
The content of conjugated linoleic acid (CLA) isomer groups in milk of two Polish sheep breeds determined by silver ion liquid chromatography (Ag(+)-HPLC).
Folia biologica (2013-06-19)
Agnieszka Rozbicka-Wieczorek, Aurelia Radzik-Rant, Witold Rant, Marian Czauderna
ABSTRACT
Sheep milk is rich in CLA isomers which are biologically active components influencing human health. There are four geometric CLA isomer pairs: cis, trans; trans, cis; trans, trans and cis, cis. The aim of the present study was the analysis of CLA isomer groups content by Ag+-HPLC in milk fat of Zelazneńska (ZS) and Wrzosówka (WS) sheep breeds. The ewes of both breeds were kept under the same environmental and nutritional conditions. Milk samples were collected from 60 suckling ewes (30 from each breed), at the age of 3-4 years and in their 4th week of lactation. A higher total amount of all CLA isomer groups was obtained in milk of ZS ewes, however, this result was statistically insignificant. The percentage of the main c9, t11 isomer in total CLA was higher in Wrzosówka milk (68% vs. 74%). The content of the trans, trans isomer group in milk fat of the studied breeds was similar. The percentage of this group in total CLA in milk of WS and ZS constitutes 7.2% and 7.7%, respectively. The amount of cis,cis isomers in milk fat of ZS was higher thanWS (P < or = 0.01).
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis