Merck
  • Environmental exposure to metals and children's growth to age 5 years: a prospective cohort study.
Environmental exposure to metals and children's growth to age 5 years: a prospective cohort study.
American journal of epidemiology (2013-05-17)
Renee M Gardner, Maria Kippler, Fahmida Tofail, Matteo Bottai, Jena Hamadani, Margaretha Grandér, Barbro Nermell, Brita Palm, Kathleen M Rasmussen, Marie Vahter
ABSTRACT
In this prospective cohort study, based on 1,505 mother-infant pairs in rural Bangladesh, we evaluated the associations between early-life exposure to arsenic, cadmium, and lead, assessed via concentrations in maternal and child urine, and children's weights and heights up to age 5 years, during the period 2001-2009. Concurrent and prenatal exposures were evaluated using linear regression analysis, while longitudinal exposure was assessed using mixed-effects linear regression. An inverse association was found between children's weight and height, age-adjusted z scores, and growth velocity at age 5 years and concurrent exposure to cadmium and arsenic. In the longitudinal analysis, multivariable-adjusted attributable differences in children's weight at age 5 years were -0.33 kg (95% confidence interval (CI): -0.60, -0.06) for high (≥95th percentile) arsenic exposure and -0.57 kg (95% CI: -0.88, -0.26) for high cadmium exposure, in comparison with children with the lowest exposure (≤5th percentile). Multivariable-adjusted attributable differences in height were -0.50 cm (95% CI: -1.20, 0.21) for high arsenic exposure and -1.6 cm (95% CI: -2.4, -0.77) for high cadmium exposure. The associations were apparent primarily among girls. The negative effects on children's growth at age 5 years attributable to arsenic and cadmium were of similar magnitude to the difference between girls and boys in terms of weight (-0.67 kg, 95% CI: -0.82, -0.53) and height (-1.3 cm, 95% CI: -1.7, -0.89).
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Arsenic, 99.999% trace metals basis
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Arsenic, powder, ≥99.997% trace metals basis