Skip to Content
Merck
All Photos(2)

Documents

396117

Sigma-Aldrich

Lead

shot, <2 mm, 99.9% trace metals basis

Synonym(s):

Lead element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

shot

reaction suitability

core: lead

resistivity

20.65 μΩ-cm

particle size

<2 mm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenyi Yuan et al.
Journal of the Air & Waste Management Association (1995), 63(4), 418-423 (2013-05-22)
A novelprocess ofmechanochemical sulfidization for the treatment of cathode ray tube (CRT)funnel glass has been investigated by co-grinding with the element sulfur The ground samples were characterized by means of a set of analytical methods, including scanning electron microscopy (SEM)
Images in clinical medicine. Pellets in the appendix.
Ibrahim Zardawi et al.
The New England journal of medicine, 369(6), e7-e7 (2013-08-09)
Kai-Ying Chiang et al.
Journal of environmental sciences (China), 25(3), 626-636 (2013-08-08)
This study investigates biogeochemical reductive release of arsenate from beudantite into solution in a crater area in northern Taiwan, using a combination of X-ray absorption near-edge structure (XANES) and atomic absorption spectrometry. Total arsenic (As) concentrations in the soil were
Plinio Carta et al.
Giornale italiano di medicina del lavoro ed ergonomia, 35(1), 32-35 (2013-06-27)
In several recent epidemiological studies blood lead levels (BLLs) even below the current CDC intervention level of 10 microg/dl have been associated with reduced neurocognitive capacities of children, with no clear evidence of a "safe" threshold. We analyzed the relationship
Karsten Witt et al.
Brain : a journal of neurology, 136(Pt 7), 2109-2119 (2013-06-27)
Deep brain stimulation of the subthalamic nucleus improves motor functions in patients suffering from advanced Parkinson's disease but in some patients, it is also associated with a mild decline in cognitive functioning about one standard deviation from the preoperative state.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service