Merck
  • The effect of robenacoxib on the concentration of C-reactive protein in synovial fluid from dogs with osteoarthritis.
The effect of robenacoxib on the concentration of C-reactive protein in synovial fluid from dogs with osteoarthritis.
BMC veterinary research (2013-03-05)
David Bennett, Peter David Eckersall, Mary Waterston, Veronica Marchetti, Alessandra Rota, Eilidh McCulloch, Silvia Sbrana
ABSTRACT
Robenacoxib is a novel and highly selective inhibitor of COX-2 in dogs and cats and because of its acidic nature is regarded as being tissue-selective. Thirty four dogs with stifle osteoarthritis secondary to failure of the cranial cruciate ligament were recruited into this study. Lameness, radiographic features, synovial cytology and C-reactive protein concentrations in serum and synovial fluid were assessed before and 28 days after commencing a course of Robenacoxib at a dose of 1 mg/kg SID. There was a significant reduction in the lameness score (P < 0.01) and an increase in the radiographic score (P < 0.05) between pre- and post-treatment assessments. There was no difference between pre- (median 1.49 mg/l; Q1-Q3 0.56-4.24 mg/L) and post - (1.10 mg/L; 0.31-1.78 mg/L) treatment serum C-reactive protein levels although synovial fluid levels were significantly reduced (pre- : 0.44 mg/L; 0.23-1.62 mg/L; post- : 0.17 mg/L; 0.05-0.49 mg/L) (P < 0.05). There was no correlation between C-reactive protein concentrations in serum and matched synovial fluid samples. Robenacoxib proved effective in reducing lameness in dogs with failure of the cranial cruciate ligament and osteoarthritis of the stifle joint. The drug also reduced levels of C-reactive protein in the synovial fluid taken from the affected stifle joint. Robenacoxib appears to reduce articular inflammation as assessed by C-reactive protein which supports the concept that Robenacoxib is a tissue-selective non-steroidal anti-inflammatory drug.
Sigma-Aldrich
Diphenylamine, ReagentPlus®, 99%
Supelco
Diphenylamine, PESTANAL®, analytical standard
Sigma-Aldrich
Diphenylamine, puriss. p.a., redox indicator, ACS reagent, reag. Ph. Eur., ≥98% (GC)
Sigma-Aldrich
Diphenylamine, ACS reagent, ≥99%
Supelco
Diphenylamine solution, certified reference material, 5000 μg/mL in methanol