Merck
  • Making bispirin: synthesis, structure and activity against Helicobacter pylori of bismuth(III) acetylsalicylate.
Making bispirin: synthesis, structure and activity against Helicobacter pylori of bismuth(III) acetylsalicylate.
Chemical communications (Cambridge, England) (2013-03-02)
Philip C Andrews, Victoria L Blair, Richard L Ferrero, Peter C Junk, Ish Kumar
ABSTRACT
Reaction of Bi(O(t)Bu)3 with aspirin (acetylsalicylic acid = aspH) in dry toluene results in the bismuth(III) complex, [Bi(O2C(C6H4)OAc)3]∞ 1 (O2C(C6H4)OAc = asp), minimum inhibitory concentration (MIC) against Helicobacter pylori ≥ 6.25 μg mL(-1), while the inclusion of a stoichiometric equivalent of KO(t)Bu leads to crystals of the bismuthate salt [KBi(O2C(C6H4)OAc)4]∞ 2.
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis