Merck
  • Modular pyridine synthesis from oximes and enals through synergistic copper/iminium catalysis.
Modular pyridine synthesis from oximes and enals through synergistic copper/iminium catalysis.
Journal of the American Chemical Society (2013-02-27)
Ye Wei, Naohiko Yoshikai
ABSTRACT
We describe here a [3+3]-type condensation reaction of O-acetyl ketoximes and α,β-unsaturated aldehydes that is synergistically catalyzed by a copper(I) salt and a secondary ammonium salt (or amine). This redox-neutral reaction allows modular synthesis of a variety of substituted pyridines under mild conditions with tolerance of a broad range of functional groups. The reaction is driven by a merger of iminium catalysis and redox activity of the copper catalyst, which would initially reduce the oxime N-O bond to generate a nucleophilic copper(II) enamide and later oxidize a dihydropyridine intermediate to the pyridine product.
Product Description
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Pyridine, ≥99%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Pyridine, ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%