Skip to Content
Merck
All Photos(1)

Key Documents

320498

Sigma-Aldrich

Pyridine

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
115 °C (lit.)
115 °C
vapor pressure:
10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

grade

reagent

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

899 °F

expl. lim.

12.4 %

dilution

(for general lab use)

refractive index

n20/D 1.509 (lit.)

pH

8.5 (25 °C, 15.82 g/L)

bp

115 °C (lit.)
115 °C

mp

−42 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Bobby Happ et al.
Chemical Society reviews, 41(6), 2222-2255 (2011-11-15)
Pyridine-based ligands, such as 2,2'-bipyridine and 1,10-phenanthroline, have gained much interest in the fields of supramolecular chemistry as well as materials science. The appealing optoelectronic properties of their complexes with heavy d(6) transition metal ions, such as Ru(ii), Os(II), Re(I)
Total syntheses of complanadines A and B.
Le Zhao et al.
Angewandte Chemie (International ed. in English), 52(6), 1722-1725 (2013-01-03)
Wen Zhou et al.
Inorganic chemistry, 52(5), 2271-2273 (2013-02-20)
The reaction of 2,2'-bipyridine (bpy) with monomeric chromium(II) precursors was used to prepare the S = 1 complexes Cr(tBu-acac)2(bpy) (1) and (η(5)-Cp)(η(1)-Cp)Cr(bpy) (3), as well as the S = 2 compound Cr[N(SiMe3)2]2(bpy) (4). The crystallographically determined bond lengths indicate that
Nathalie Ségaud et al.
Inorganic chemistry, 52(2), 691-700 (2013-01-11)
We report the synthesis, characterization, and solution chemistry of a series of new Fe(II) complexes based on the tetradentate ligand N-methyl-N,N'-bis(2-pyridyl-methyl)-1,2-diaminoethane or the pentadentate ones N,N',N'-tris(2-pyridyl-methyl)-1,2-diaminoethane and N,N',N'-tris(2-pyridyl-methyl)-1,3-diaminopropane, modified by propynyl or methoxyphenyltriazolyl groups on the amino functions. Six of
Maria Milina et al.
Angewandte Chemie (International ed. in English), 54(5), 1591-1594 (2014-11-08)
Without techniques sensitive to complex pore architectures, synthetic efforts to enhance molecular transport in zeolite and other porous materials through hierarchical structuring lack descriptors for their rational design. The power of positron annihilation lifetime spectroscopy (PALS) to characterize the pore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service