Merck
  • Evaluation of a novel chitosan polymer-based adsorbent for the removal of chromium (III) in aqueous solutions.
Evaluation of a novel chitosan polymer-based adsorbent for the removal of chromium (III) in aqueous solutions.
Carbohydrate polymers (2013-02-13)
XiaoJun Zuo, Rajasekhar Balasubramanian
ABSTRACT
Novel poly(vinyl alcohol)/citric acid/chitosan (PVA/CA/CHT, PCC) beads were prepared as an adsorbent for the removal of trivalent chromium (Cr(3+)) in aqueous solutions. PCC beads with different mass ratios (I, II, III, IV, V, ad VI) between PVA/CA/CHT were evaluated for the removal of Cr(3+) ions to find an optimal stoichiometry of PCC beads, which indicated that the PCC bead (II) with the PVA/CA/CHT mass ratio of 1/5/1 showed the highest adsorption for Cr(3+) ions. The surface characteristics of PCC beads (II) were determined by scanning electron microscopy using a field emission scanning electron microscope (FESEM) and Fourier transform infrared spectroscopy (FTIR). The results obtained in this study implied that PCC bead (II) had an obviously rough and folded structure, and its spectrum manifested significant changes compared to those of PVA, CA and CHT. The sorption experiments were conducted to evaluate the performance of PCC beads (II) to adsorb Cr(3+) ions in aqueous solutions. Results indicated that the Cr(3+) ion sorption onto PCC beads (II) was highly pH-dependent with the maximum uptake at pH 6.0. The sorption kinetics data were well fitted by the pseudo-second-order equation with high regression coefficients. Sorption isotherm of PCC beads (II) was well described by Langmuir equation, with a maximum sorption at 41.5mg Cr/g PCC beads (II). Desorption studies were also carried out by the repeated sorption/desorption experiments. The high efficiency and reusability of PCC beads (II) make them an attractive sorbent for the removal of Cr(3+) and possibly other metals in aqueous solutions.
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000