Merck
  • Outcomes of patients with microscopic colitis treated with corticosteroids: a population-based study.
Outcomes of patients with microscopic colitis treated with corticosteroids: a population-based study.
The American journal of gastroenterology (2013-01-09)
Nicole M Gentile, Adil A Abdalla, Sahil Khanna, Thomas C Smyrk, William J Tremaine, William A Faubion, Patricia P Kammer, William J Sandborn, Edward V Loftus, Darrell S Pardi
ABSTRACT
To evaluate the outcomes of corticosteroid-treated microscopic colitis (MC) in a population-based cohort, and to compare these outcomes in patients treated with prednisone or budesonide. A historical cohort study of Olmsted County, Minnesota residents diagnosed with collagenous or lymphocytic colitis (LC) between 1986 and 2010 was performed using the Rochester Epidemiology Project. Of 315 patients with MC, 80 (25.4%) were treated with corticosteroids. The median age at colitis diagnosis was 66.5 years (range: 16-95) and 78.7% were female. Forty patients (50%) had LC and 40 (50%) had collagenous colitis. Prednisone was used in 17 patients (21.2%) and budesonide in 63 (78.8%); 56 (75.6%) had complete response and 15 (20.3%) had partial response. Patients treated with budesonide had a higher rate of complete response than those treated with prednisone (82.5 vs. 52.9%; odds ratio, 4.18; 95% CI, 1.3-13.5). Six patients were lost to follow-up. The remaining 74 had a median follow-up of 4 years (range 0.2-14). Fifty patients out of the 71 who responded (70.4%) had a recurrence after corticosteroid discontinuation. Patients treated with budesonide were less likely to recur than those treated with prednisone (hazard ratio, 0.38; 95% CI, 0.18-0.85; P=0.02). After 397 person years of follow-up in the 73 patients with long-term data, 47 (64.4%) required maintenance with corticosteroids. Patients with MC often respond to corticosteroid therapy, but with a high relapse rate. Budesonide had a higher response rate and a lower risk of recurrence than prednisone.
Sigma-Aldrich
Budesonide, ≥99%
Budesonide, European Pharmacopoeia (EP) Reference Standard
Supelco
Budesonide, Pharmaceutical Secondary Standard; Certified Reference Material