Merck
  • Methyl eugenol: its occurrence, distribution, and role in nature, especially in relation to insect behavior and pollination.
Methyl eugenol: its occurrence, distribution, and role in nature, especially in relation to insect behavior and pollination.
Journal of insect science (Online) (2012-09-12)
Keng Hong Tan, Ritsuo Nishida
ABSTRACT
This review discusses the occurrence and distribution (within a plant) of methyl eugenol in different plant species (> 450) from 80 families spanning many plant orders, as well as various roles this chemical plays in nature, especially in the interactions between tephritid fruit flies and plants.
Product Description
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
4-Allyl-1,2-dimethoxybenzene, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Methyl eugenol, ≥98%, FCC
Supelco
Eugenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Methyl eugenol, analytical standard
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Eugenol, European Pharmacopoeia (EP) Reference Standard