Merck
  • Toward rational design of amine solutions for PCC applications: the kinetics of the reaction of CO2(aq) with cyclic and secondary amines in aqueous solution.
Toward rational design of amine solutions for PCC applications: the kinetics of the reaction of CO2(aq) with cyclic and secondary amines in aqueous solution.
Environmental science & technology (2012-05-25)
William Conway, Xiaoguang Wang, Debra Fernandes, Robert Burns, Geoffrey Lawrance, Graeme Puxty, Marcel Maeder
ABSTRACT
The kinetics of the fast reversible carbamate formation reaction of CO(2)(aq) with a series of substituted cyclic secondary amines as well as the noncyclic secondary amine diethanolamine (DEA) has been investigated using the stopped-flow spectrophotometric technique at 25.0 °C. The kinetics of the slow parallel reversible reaction between HCO(3)(-) and amine has also been determined for a number of the amines by (1)H NMR spectroscopy at 25.0 °C. The rate of the reversible reactions and the equilibrium constants for the formation of carbamic acid/carbamate from the reactions of CO(2) and HCO(3)(-) with the amines are reported. In terms of the forward reaction of CO(2)(aq) with amine, the order with increasing rate constants is as follows: diethanolamine (DEA) < morpholine (MORP) ~ thiomorpholine (TMORP) < N-methylpiperazine (N-MPIPZ) < 4-piperidinemethanol (4-PIPDM) ~ piperidine (PIPD) < pyrrolidine (PYR). Both 2-piperidinemethanol (2-PIPDM) and 2-piperidineethanol (2-PIPDE) do not form carbamates. For the carbamate forming amines a Brønsted correlation relating the protonation constant of the amine to the carbamic acid formation rate and equilibrium constants at 25.0 °C has been established. The overall suitability of an amine for PCC in terms of kinetics and energy is discussed.
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Supelco
Diethanolamine, analytical standard
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Diethanolamine hydrochloride, 98%