Skip to Content
Merck
All Photos(1)

Key Documents

31590

Sigma-Aldrich

Diethanolamine

puriss. p.a., ACS reagent, ≥99.0% (GC)

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
217 °C/150 mmHg (lit.)
vapor pressure:
<0.98 atm ( 100 °C)
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

Assay

≥99.0% (GC)

form

liquid
viscous liquid

autoignition temp.

689 °F

expl. lim.

10.6 %

technique(s)

GC/GC: suitable

impurities

≤0.15% water

refractive index

n20/D 1.477 (lit.)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤1 mg/kg

functional group

amine
hydroxyl

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethanolamine (DEA) is an alkanolamine. Kinetic studies of its reaction with carbon dioxide has been proposed.[1]

Application

Diethanolamine (DEA) may be used to compose the Ti(O-i-Pr)4 (titanium isopropoxide)-diethanolamine-i-propanol system, required for the preparation of transparent TiO2 films by dip-coating method.[2] It may be used for the coating of ZnO nanoparticles.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dip-coating of TiO2 films using a sol derived from Ti(Oi-Pr)4-diethanolamine-H2O-i-PrOH system.
Takahashi Y and Matsuoka Y.
J. Mater. Sci., 23(6), 2259-2266 (1988)
Synthesis of ZnO nanoparticles with tunable emission colors and their cell labeling applications.
Tang X, et al.
Chemistry of Materials, 22(11), 3383-3388 (2010)
The reaction between CO2 and diethanolamine at 298 K.
Versteeg GF and Oyevaar MH.
Chemical Engineering Science, 44(5), 1264-1268 (1989)
Bjørn Henrik Hansen et al.
Aquatic toxicology (Amsterdam, Netherlands), 99(2), 212-222 (2010-06-12)
Alkanolamines are surface-active chemicals used in a wide range of industrial, agricultural and pharmaceutical applications and products. Of particular interest is the use of alkanolamines such as diethanolamine (DEA) in the removal of CO(2) from natural gas and for CO(2)
Wenting Zhang et al.
PloS one, 10(5), e0125995-e0125995 (2015-05-15)
We previously reported a new approach for culturing difficult-to-preserve primary patient-derived multiple myeloma cells (MMC) using an osteoblast (OSB)-derived 3D tissue scaffold constructed in a perfused microfluidic environment and a culture medium supplemented with patient plasma. In the current study

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service