Merck
  • A fluorescence-based microtiter plate assay for γ-glutamylcyclotransferase.
A fluorescence-based microtiter plate assay for γ-glutamylcyclotransferase.
Analytical biochemistry (2011-10-18)
Philip G Board, Inca Hutchinson
ABSTRACT
γ-Glutamylcyclotransferase (GGCT) is a component of the γ-glutamyl cycle and specifically cleaves γ-glutamyl-amino acid dipeptides to release the free amino acid and 5-oxoproline. The action of GGCT in glutathione synthetase-deficient patients results in the accumulation of high levels of 5-oxoproline. In addition, GGCT may be a biomarker in some cancers. GGCT inhibitors, therefore, may be of value in the treatment of glutathione synthetase deficiency or in the elucidation of the role of GGCT in cancer cells. Published GGCT assays are not suitable for high-throughput screening for inhibitory molecules. We have developed a fluorescence-based 96-well plate assay for the determination of the rate of γ-glutamylcysteine cleavage by GGCT. After the reaction, the residual γ-glutamylcysteine is determined fluorometrically after derivatization with 2,3-naphthalenedicarboxaldehyde. The method has sufficient sensitivity to detect low-affinity competitive inhibitors and, as a result of its simplicity and microtiter plate format, can be readily used in high-throughput inhibitor screens.
Product Description
Supelco
Sodium tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]borate trihydrate, Selectophore
Sigma-Aldrich
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence
Sigma-Aldrich
γ-Glu-Cys, ≥80% (HPLC)