Merck
  • Discovery of WRN inhibitor HRO761 with synthetic lethality in MSI cancers.
Discovery of WRN inhibitor HRO761 with synthetic lethality in MSI cancers.
Nature (2024-04-25)
Stephane Ferretti, Jacques Hamon, Ruben de Kanter, Clemens Scheufler, Rita Andraos-Rey, Stephanie Barbe, Elisabeth Bechter, Jutta Blank, Vincent Bordas, Ernesta Dammassa, Andrea Decker, Noemi Di Nanni, Marion Dourdoigne, Elena Gavioli, Marc Hattenberger, Alisa Heuser, Christelle Hemmerlin, Jürgen Hinrichs, Grainne Kerr, Laurent Laborde, Isabel Jaco, Eloísa Jiménez Núñez, Hans-Joerg Martus, Cornelia Quadt, Markus Reschke, Vincent Romanet, Fanny Schaeffer, Joseph Schoepfer, Maxime Schrapp, Ross Strang, Hans Voshol, Markus Wartmann, Sarah Welly, Frédéric Zécri, Francesco Hofmann, Henrik Möbitz, Marta Cortés-Cros
ABSTRACT
The Werner syndrome RecQ helicase WRN was identified as a synthetic lethal target in cancer cells with microsatellite instability (MSI) by several genetic screens1-6. Despite advances in treatment with immune checkpoint inhibitors7-10, there is an unmet need in the treatment of MSI cancers11-14. Here we report the structural, biochemical, cellular and pharmacological characterization of the clinical-stage WRN helicase inhibitor HRO761, which was identified through an innovative hit-finding and lead-optimization strategy. HRO761 is a potent, selective, allosteric WRN inhibitor that binds at the interface of the D1 and D2 helicase domains, locking WRN in an inactive conformation. Pharmacological inhibition by HRO761 recapitulated the phenotype observed by WRN genetic suppression, leading to DNA damage and inhibition of tumour cell growth selectively in MSI cells in a p53-independent manner. Moreover, HRO761 led to WRN degradation in MSI cells but not in microsatellite-stable cells. Oral treatment with HRO761 resulted in dose-dependent in vivo DNA damage induction and tumour growth inhibition in MSI cell- and patient-derived xenograft models. These findings represent preclinical pharmacological validation of WRN as a therapeutic target in MSI cancers. A clinical trial with HRO761 (NCT05838768) is ongoing to assess the safety, tolerability and preliminary anti-tumour activity in patients with MSI colorectal cancer and other MSI solid tumours.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1), liquid, clone DO-1, Calbiochem®
Sigma-Aldrich
Anti-MDM2 (Ab-1) Mouse mAb (IF2), liquid, clone IF2, Calbiochem®
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Monoclonal Anti-Cyclin A antibody produced in mouse, clone CY-A1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Werner′s syndrome helicase (WRN) Antibody, clone 20A11.1, clone 20A11.1, from mouse
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®