Merck
  • The First Transcriptomic Atlas of the Adult Lacrimal Gland Reveals Epithelial Complexity and Identifies Novel Progenitor Cells in Mice.
The First Transcriptomic Atlas of the Adult Lacrimal Gland Reveals Epithelial Complexity and Identifies Novel Progenitor Cells in Mice.
Cells (2023-07-06)
Vanessa Delcroix, Olivier Mauduit, Hyun Soo Lee, Anastasiia Ivanova, Takeshi Umazume, Sarah M Knox, Cintia S de Paiva, Darlene A Dartt, Helen P Makarenkova
ABSTRACT
The lacrimal gland (LG) secretes aqueous tears. Previous studies have provided insights into the cell lineage relationships during tissue morphogenesis. However, little is known about the cell types composing the adult LG and their progenitors. Using scRNAseq, we established the first comprehensive cell atlas of the adult mouse LG to investigate the cell hierarchy, its secretory repertoire, and the sex differences. Our analysis uncovered the complexity of the stromal landscape. Epithelium subclustering revealed myoepithelial cells, acinar subsets, and two novel acinar subpopulations: Tfrchi and Car6hi cells. The ductal compartment contained Wfdc2+ multilayered ducts and an Ltf+ cluster formed by luminal and intercalated duct cells. Kit+ progenitors were identified as: Krt14+ basal ductal cells, Aldh1a1+ cells of Ltf+ ducts, and Sox10+ cells of the Car6hi acinar and Ltf+ epithelial clusters. Lineage tracing experiments revealed that the Sox10+ adult populations contribute to the myoepithelial, acinar, and ductal lineages. Using scRNAseq data, we found that the postnatally developing LG epithelium harbored key features of putative adult progenitors. Finally, we showed that acinar cells produce most of the sex-biased lipocalins and secretoglobins detected in mouse tears. Our study provides a wealth of new data on LG maintenance and identifies the cellular origin of sex-biased tear components.
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Anti-Keratin 14 antibody, Rabbit monoclonal, clone SP53, recombinant, expressed in proprietary host, affinity isolated antibody
Sigma-Aldrich
Anti-Heparan Sulfate Proteoglycan (Perlecan) Antibody, clone A7L6, clone A7L6, from rat