Merck
  • Inhibition of protein glycosylation is a novel pro-angiogenic strategy that acts via activation of stress pathways.
Inhibition of protein glycosylation is a novel pro-angiogenic strategy that acts via activation of stress pathways.
Nature communications (2020-12-12)
Cuiling Zhong, Pin Li, Sulabha Argade, Lixian Liu, Anastasia Chilla', Wei Liang, Hong Xin, Brian Eliceiri, Biswa Choudhury, Napoleone Ferrara
ABSTRACT
Endothelial cell (EC) metabolism is thought to be one of the driving forces for angiogenesis. Here we report the identification of the hexosamine D-mannosamine (ManN) as an EC mitogen and survival factor for bovine and human microvascular EC, with an additivity with VEGF. ManN inhibits glycosylation in ECs and induces significant changes in N-glycan and O-glycan profiles. We further demonstrate that ManN and two N-glycosylation inhibitors stimulate EC proliferation via both JNK activation and the unfolded protein response caused by ER stress. ManN results in enhanced angiogenesis in a mouse skin injury model. ManN also promotes angiogenesis in a mouse hindlimb ischemia model, with accelerated limb blood flow recovery compared to controls. In addition, intraocular injection of ManN induces retinal neovascularization. Therefore, activation of stress pathways following inhibition of protein glycosylation can promote EC proliferation and angiogenesis and may represent a therapeutic strategy for treatment of ischemic disorders.
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK8
Sigma-Aldrich
α-Mannosidase from Canavalia ensiformis (Jack bean), ammonium sulfate suspension, ≥15 units/mg protein (biuret)
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized powder, ≥2 units/mg solid
Sigma-Aldrich
Keratinocyte Growth Factor human, KGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
4-Phenylbutyric acid, 99%
Sigma-Aldrich
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
USP
Mannose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
4-Nitrophenyl α-D-glucopyranoside, ≥99%
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
D-Mannosamine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Kifunensine, mannosidase inhibitor
USP
Glucosamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Mass Spectrometry Metabolite Library, Supplied by IROA Technologies
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK9
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%