Merck
  • The role of amphiphilicity and negative charge in glycoprotein 41 interactions in the hydrophobic pocket.
The role of amphiphilicity and negative charge in glycoprotein 41 interactions in the hydrophobic pocket.
Journal of medicinal chemistry (2009-06-19)
Miriam Gochin, Lifeng Cai
ABSTRACT
The hydrophobic pocket within the coiled coil domain of HIV-1 gp41 is considered to be a hot-spot suitable for small molecule intervention of fusion, although so far it has yielded only microM inhibitors. Previous peptide studies have identified specific hydrophobic interactions and a Lys-Asp salt bridge as contributing to binding affinity in the pocket. Negative charge appears to be critical for activity of small molecules. We have examined the role of charge and amphiphilic character in the interaction by studying a series of short pocket binding peptides differing in charge, helical content, and in the presence or absence of the Lys-Asp salt bridge, and a series of fatty acid salts with varying charge and hydrocarbon length. Quantitative binding analysis revealed that long-range electrostatic forces and a greasy nonspecific hydrophobic interaction were sufficient for microM potency. The results suggest that an extended interaction site may be necessary for higher potency. We examined a region of the coiled coil immediately C-terminal to the pocket and found that specific salt bridge and hydrogen bond networks may reside in this region. Negatively charged groups extended toward or beyond the C-terminus of the pocket could therefore result in improved low molecular weight fusion inhibitors.
Sigma-Aldrich
Tetradecyl sulfate sodium salt, 95%
Sigma-Aldrich
Sodium 1-dodecanesulfonate, ReagentPlus®, ≥99%
Supelco
Sodium octyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
Sodium 1-decanesulfonate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecanoate, 99-100%
Sigma-Aldrich
Sodium octyl sulfate, ≥95%
Sigma-Aldrich
Sodium 1-decanesulfonate, ~98%
Sigma-Aldrich
Sodium hexanesulfonate, ≥98% (elemental analysis)
Sigma-Aldrich
Sodium hexanesulfonate, BioXtra
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium 1-decanesulfonate, ≥99.0% (T)
Sigma-Aldrich
Octadecyl sulfate sodium salt, technical grade, 93%
Sigma-Aldrich
Sodium octanoate, ≥99% (capillary GC)
Supelco
Sodium decyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
1-Hexadecanesulfonic acid sodium salt, 98%