Merck
  • The Ig superfamily protein PTGFRN coordinates survival signaling in glioblastoma multiforme.
The Ig superfamily protein PTGFRN coordinates survival signaling in glioblastoma multiforme.
Cancer letters (2019-08-05)
Brittany Aguila, Adina Brett Morris, Raffaella Spina, Eli Bar, Julie Schraner, Robert Vinkler, Jason W Sohn, Scott M Welford
ABSTRACT
Glioblastoma multiforme (GBM) is the most malignant primary brain tumor with a median survival of approximately 14 months. Despite aggressive treatment of surgical resection, chemotherapy and radiation therapy, only 3-5% of GBM patients survive more than 3 years. Contributing to this poor therapeutic response, it is believed that GBM contains both intrinsic and acquired mechanisms of resistance, including resistance to radiation therapy. In order to define novel mediators of radiation resistance, we conducted a functional knockdown screen, and identified the immunoglobulin superfamily protein, PTGFRN. In GBM, PTGFRN is found to be overexpressed and to correlate with poor survival. Reducing PTGFRN expression radiosensitizes GBM cells and potently decreases the rate of cell proliferation and tumor growth. Further, PTGFRN inhibition results in significant reduction of PI3K p110β and phosphorylated AKT, due to instability of p110β. Additionally, PTGFRN inhibition decreases nuclear p110β leading to decreased DNA damage sensing and DNA damage repair. Therefore overexpression of PTGFRN in glioblastoma promotes AKT-driven survival signaling and tumor growth, as well as increased DNA repair signaling. These findings suggest PTGFRN is a potential signaling hub for aggressiveness in GBM.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-CD9 antibody produced in mouse, clone 4A2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-PIK3CB antibody produced in mouse, clone 10D5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-CD9 antibody produced in rabbit, affinity isolated antibody