Merck
  • Clinical significance of gelsolin-like actin-capping protein expression in oral carcinogenesis: an immunohistochemical study of premalignant and malignant lesions of the oral cavity.
Clinical significance of gelsolin-like actin-capping protein expression in oral carcinogenesis: an immunohistochemical study of premalignant and malignant lesions of the oral cavity.
BMC cancer (2008-02-02)
Hitomi Nomura, Katsuhiro Uzawa, Takashi Ishigami, Yukinao Kouzu, Hirofumi Koike, Katsunori Ogawara, Masashi Siiba, Hiroki Bukawa, Hidetaka Yokoe, Hitoshi Kubosawa, Hideki Tanzawa
ABSTRACT
Gelsolin-like actin-capping protein (CapG) is a ubiquitous gelsolin-family actin-modulating protein involved in cell signalling, receptor-mediated membrane ruffling, phagocytosis, and motility. CapG has generated great interest due to its oncogenic function in the control of cell migration or invasion in a variety of cancer cells. We previously applied proteomic methods to characterize differentially expressed proteins in oral squamous-cell carcinoma (OSCC) cells and detected significantly high expression levels of CapG in OSCC-derived cell lines compared to human normal oral keratinocytes. In the current study, to further determine the potential involvement of CapG in OSCC, we evaluated the status of CapG protein and mRNA expression in human oral premalignant lesions (OPLs) and primary OSCCs and correlated the results with clinicopathologic variables. Matched normal and tumour tissue sections of 79 human primary OSCCs and 28 OPLs were analyzed for CapG expression by immunohistochemistry (IHC). Correlations between CapG-immunohistochemical staining scores of OSCCs and clinicopathologic features were evaluated by Fisher's exact test. Real-time quantitative reverse transcriptase-polymerase chain reaction (qRT-PCR) was used to estimate CapG expression at the mRNA level. In IHC, substantial up-regulation of CapG protein was observed in primary OSCCs (52%) and OPLs (64%), whereas corresponding normal tissues showed consistently weak or absent immunoreactivity of CapG. qRT-PCR data were consistent with the protein expression status. Moreover, CapG expression was correlated with the TNM stage grading of OSCCs. Our finding of frequent dysregulated expression of CapG in premalignant and malignant lesions together with an association with an advanced clinical disease stage suggests that CapG could contribute to cancer development and progression and that CapG may have potential as a biomarker and a therapeutic target for OSCC.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and sodium bicarbonate, without HEPES, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and trace elements, without HEPES and sodium bicarbonate, powder, suitable for cell culture
SAFC
DMEM/Nutrient Mixture F-12 Ham, With 15 mM HEPES, without L-glutamine, L-leucine, L-lysine, L-methionine, CaCl2, MgCl2, MgSO4, sodium bicarbonate, and phenol red, powder, suitable for cell culture
Sigma-Aldrich
StableCell DMEM/F12, With stable glutamine, 15mM HEPES and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture