Merck
  • RLN3/RXFP3 Signaling in the PVN Inhibits Magnocellular Neurons via M-like Current Activation and Contributes to Binge Eating Behavior.
RLN3/RXFP3 Signaling in the PVN Inhibits Magnocellular Neurons via M-like Current Activation and Contributes to Binge Eating Behavior.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2020-06-14)
Alan Kania, Agata Szlaga, Patryk Sambak, Anna Gugula, Ewa Blasiak, Maria Vittoria Micioni Di Bonaventura, Mohammad Akhter Hossain, Carlo Cifani, Grzegorz Hess, Andrew L Gundlach, Anna Blasiak
ABSTRACT
Binge-eating disorder is the most common eating disorder. Various neuropeptides play important roles in the regulation of feeding behavior, including relaxin-3 (RLN3), which stimulates food intake in rats through the activation of the relaxin-family peptide-3 receptor (RXFP3). Here we demonstrate that a likely mechanism underlying the orexigenic action of RLN3 is RXFP3-mediated inhibition of oxytocin- and arginine-vasopressin-synthesizing paraventricular nucleus (PVN) magnocellular neurosecretory cells. Moreover, we reveal that, in male and female rats, this action depends on M-like potassium conductance. Notably, higher intra- and peri-PVN RLN3 fiber densities were observed in females, which may constitute an anatomic substrate for observed sex differences in binge-eating disorder. Finally, in a model of binge-eating in female rats, RXFP3 blockade within the PVN prevented binge-eating behavior. These data demonstrate a direct RLN3/RXFP3 action in the PVN of male and female rats, identify the associated ionic mechanisms, and reveal that hypothalamic RLN3/RXFP3 signaling regulates binge-eating behavior.SIGNIFICANCE STATEMENT Binge-eating disorder is the most common eating disorder worldwide, affecting women twice as frequently as men. Various neuropeptides play important roles in the regulation of feeding behavior, including relaxin-3, which acts via the relaxin-family peptide-3 receptor (RXFP3). Using a model of binge-eating, we demonstrated that relaxin-3/RXFP3 signaling in the hypothalamic paraventricular nucleus (PVN) is necessary for the expression of binge-eating behavior in female rats. Moreover, we elucidated the neuronal mechanism of RLN3/RXFP3 signaling in PVN in male and female rats and characterized sex differences in the RLN3 innervation of the PVN. These findings increase our understanding of the brain circuits and neurotransmitters involved in binge-eating disorder pathology and identify RXFP3 as a therapeutic target for binge-like eating disorders.
Sigma-Aldrich
Fluoroshield with DAPI, liquid
Pricing and availability is not currently available.
Sigma-Aldrich
Cadmium chloride hydrate, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Cadmium chloride hydrate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Oxytocin Antibody, clone 4G11, ascites fluid, clone 4G11, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methiodide, ≥95.0% (HPCE)
Pricing and availability is not currently available.
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Pricing and availability is not currently available.