Skip to Content
Merck
All Photos(3)

Key Documents

B4261

Sigma-Aldrich

Biocytin

≥98% (TLC), suitable for drug transporter assays

Synonym(s):

Bct, Nε-(+)-Biotinyl-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28N4O4S
CAS Number:
Molecular Weight:
372.48
Beilstein:
97197
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Biocytin, ≥98% (TLC)

Assay

≥98% (TLC)

form

powder

optical activity

[α]20/D +53°, c = 1.05% in 0.1 M NaOH(lit.)

technique(s)

drug transporter assay: suitable

color

white to off-white

mp

~245 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](N)C(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C16H28N4O4S/c17-10(15(22)23)5-3-4-8-18-13(21)7-2-1-6-12-14-11(9-25-12)19-16(24)20-14/h10-12,14H,1-9,17H2,(H,18,21)(H,22,23)(H2,19,20,24)/t10-,11-,12-,14-/m0/s1

InChI key

BAQMYDQNMFBZNA-MNXVOIDGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biocytin is used as a substrate to study the specificity and kinetics of biotinidase(s), to measure biotinidase deficiency and as a model compound to study various uptake mechanisms of cells and tissues. Biocytin has been used to label neurons, medium spiny neurons and optic nerves.

Biochem/physiol Actions

Biocytin is an adduct of lysine and biotin. It is hydrolyzed by biotinidase, which acts on peptide-incorporated biotin or free biotin. Biocytin acts as a coenzyme.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Tian et al.
Theranostics, 8(3), 735-745 (2018-01-19)
Purpose: Radionuclide therapy directed against tumors that express somatostatin receptors (SSTRs) has proven effective for the treatment of advanced, low- to intermediate-grade neuroendocrine tumors in the clinic. In clinical usage, somatostatin peptide-based analogs, labeled with therapeutic radionuclides, provide an overall
Vishwa Mohan et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(32), 6233-6250 (2019-06-12)
Dendritic spines in the developing mammalian neocortex are initially overproduced and then eliminated during adolescence to achieve appropriate levels of excitation in mature networks. We show here that the L1 family cell adhesion molecule Close Homolog of L1 (CHL1) and
Jeanne T Paz et al.
Nature neuroscience, 16(1), 64-70 (2012-11-13)
Cerebrocortical injuries such as stroke are a major source of disability. Maladaptive consequences can result from post-injury local reorganization of cortical circuits. For example, epilepsy is a common sequela of cortical stroke, but the mechanisms responsible for seizures following cortical
Treadmill exercise reverses dendritic spine loss in direct and indirect striatal medium spiny neurons in the 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) mouse model of Parkinson?s disease
William A
Neurobiology of Disease (2014)
Cytometry (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service