Merck
  • TMIE Defines Pore and Gating Properties of the Mechanotransduction Channel of Mammalian Cochlear Hair Cells.
TMIE Defines Pore and Gating Properties of the Mechanotransduction Channel of Mammalian Cochlear Hair Cells.
Neuron (2020-04-29)
Christopher L Cunningham, Xufeng Qiu, Zizhen Wu, Bo Zhao, Guihong Peng, Ye-Hyun Kim, Amanda Lauer, Ulrich Müller
ABSTRACT
TMC1 and TMC2 (TMC1/2) have been proposed to form the pore of the mechanotransduction channel of cochlear hair cells. Here, we show that TMC1/2 cannot form mechanotransduction channels in cochlear hair cells without TMIE. TMIE binds to TMC1/2, and a TMIE mutation that perturbs TMC1/2 binding abolishes mechanotransduction. N-terminal TMIE deletions affect the response of the mechanotransduction channel to mechanical force. Similar to mechanically gated TREK channels, the C-terminal cytoplasmic TMIE domain contains charged amino acids that mediate binding to phospholipids, including PIP2. TMIE point mutations in the C terminus that are linked to deafness disrupt phospholipid binding, sensitize the channel to PIP2 depletion from hair cells, and alter the channel's unitary conductance and ion selectivity. We conclude that TMIE is a subunit of the cochlear mechanotransduction channel and that channel function is regulated by a phospholipid-sensing domain in TMIE with similarity to those in other mechanically gated ion channels.
Product Description
Millipore
EZview Red Anti-HA Affinity Gel
Sigma-Aldrich
1,2-Bis(2-Aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid, 98%
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-TMIE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
p-Phenylenediamine, 98% (GC)
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid